CAS RN 7355-58-0
IUPAC Name:
N-(2-chloroethyl)acetamide
Synonyms:
N-(2-chloroethyl)acetamide
Molecular Weight (g/mol):
121.56
Molecular Formula:
C4H8ClNO
InChi Key:
HSKNJSHFPPHTAQ-UHFFFAOYSA-N
SMILES:
CC(=O)NCCCl
1
–
1
of
1
results
1
N-(2-Chloroethyl)acetamide, 98%
CAS: 7355-58-0 Molecular Formula: C4H8ClNO Molecular Weight (g/mol): 121.57 MDL Number: MFCD00000965 InChI Key: HSKNJSHFPPHTAQ-UHFFFAOYSA-N Synonym: n-2-chloroethyl acetamide,acetamide, n-2-chloroethyl,n-2-chloro-ethyl-acetamide,n-acetyl-2-chloroethylamine,n 2-chloroethyl acetamide,n-acetyl-ss-chlorathylamin,acmc-1bi05,n-2-chloroethyl-acetamide,4-04-00-00449 beilstein handbook reference PubChem CID: 81815 IUPAC Name: N-(2-chloroethyl)acetamide SMILES: CC(=O)NCCCl