accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 73583-37-6

ClBrN

CAS RN 73583-37-6

IUPAC Name: 4-bromo-2-chloropyridine
Synonyms: 4-bromo-2-chloropyridine
Molecular Weight (g/mol): 192.44
Molecular Formula: C5H3BrClN
InChi Key: ONHMWUXYIFULDO-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=CC=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)