accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SiClClFFFFFFFFFFFFF

CAS RN 73609-36-6

IUPAC Name: dichloro(methyl)(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Synonyms: dichloro(methyl)(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Molecular Weight (g/mol): 461.12
Molecular Formula: C9H7Cl2F13Si
InChi Key: VBDMVWQNRXVEGC-UHFFFAOYSA-N
SMILES: C[Si](Cl)(Cl)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results