CAS RN 7365-82-4
Thermo Scientific Chemicals ACES, 99+%, for biochemistry
CAS: 7365-82-4 Molecular Formula: C4H10N2O4S Molecular Weight (g/mol): 182.194 MDL Number: MFCD00008030 InChI Key: DBXNUXBLKRLWFA-UHFFFAOYSA-N Synonym: N-(2-Acetamido)-2-aminoethanesulfonic acid,N-(Carbamoylmethyl)taurine PubChem CID: 81832 ChEBI: CHEBI:39062 IUPAC Name: 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid SMILES: C(CS(=O)(=O)O)NCC(=O)N
ACES, 0.5M buffer soln., pH 6.5, Thermo Scientific™
CAS: 7365-82-4 Molecular Formula: C4H10N2O4S Molecular Weight (g/mol): 182.194 MDL Number: MFCD00008030 InChI Key: DBXNUXBLKRLWFA-UHFFFAOYSA-N PubChem CID: 81832 ChEBI: CHEBI:39062 IUPAC Name: 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid SMILES: C(CS(=O)(=O)O)NCC(=O)N
Thermo Scientific Chemicals ACES, 99%, extra pure
CAS: 7365-82-4 Molecular Formula: C4H10N2O4S Molecular Weight (g/mol): 182.194 MDL Number: MFCD00008030 InChI Key: DBXNUXBLKRLWFA-UHFFFAOYSA-N Synonym: N-(2-Acetamido)-2-aminoethanesulfonic acid,N-(Carbamoylmethyl)taurine PubChem CID: 81832 ChEBI: CHEBI:39062 IUPAC Name: 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid SMILES: C(CS(=O)(=O)O)NCC(=O)N
Thermo Scientific Chemicals ACES Buffer (0.5 M, pH 7.0)
CAS: 7365-82-4 Molecular Formula: C4H10N2O4S Molecular Weight (g/mol): 182.194 MDL Number: MFCD00008030 InChI Key: DBXNUXBLKRLWFA-UHFFFAOYSA-N PubChem CID: 81832 ChEBI: CHEBI:39062 IUPAC Name: 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid SMILES: C(CS(=O)(=O)O)NCC(=O)N
Thermo Scientific Chemicals ACES, 99%
CAS: 7365-82-4 Molecular Formula: C4H10N2O4S Molecular Weight (g/mol): 182.194 MDL Number: MFCD00008030 InChI Key: DBXNUXBLKRLWFA-UHFFFAOYSA-N Synonym: aces,n-carbamoylmethyl taurine,n-2-acetamido-2-aminoethanesulfonic acid,2-2-amino-2-oxoethyl amino ethanesulfonic acid,ethanesulfonic acid, 2-2-amino-2-oxoethyl amino,unii-i5spl5yvby,i5spl5yvby,2-carbamoylmethylamino ethanesulfonic acid PubChem CID: 81832 ChEBI: CHEBI:39062 IUPAC Name: 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid SMILES: C(CS(=O)(=O)O)NCC(=O)N
Melford ACES
Zwitterionic buffer used in biochemistry and molecular biology research.
ACES, 0.5M buffer soln., pH 6.0, Thermo Scientific™
CAS: 7365-82-4 Molecular Formula: C4H10N2O4S Molecular Weight (g/mol): 182.194 MDL Number: MFCD00008030 InChI Key: DBXNUXBLKRLWFA-UHFFFAOYSA-N PubChem CID: 81832 ChEBI: CHEBI:39062 IUPAC Name: 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid SMILES: C(CS(=O)(=O)O)NCC(=O)N
ACES, 0.5M buffer soln., pH 7.5, Thermo Scientific™
CAS: 7365-82-4 Molecular Formula: C4H10N2O4S Molecular Weight (g/mol): 182.194 MDL Number: MFCD00008030 InChI Key: DBXNUXBLKRLWFA-UHFFFAOYSA-N PubChem CID: 81832 ChEBI: CHEBI:39062 IUPAC Name: 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid SMILES: C(CS(=O)(=O)O)NCC(=O)N