accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7366-56-5

NH2SNHOHO

CAS RN 7366-56-5

IUPAC Name: 4-(carbamothioylamino)benzoic acid
Synonyms: 4-(carbamothioylamino)benzoic acid
Molecular Weight (g/mol): 196.22
Molecular Formula: C8H8N2O2S
InChi Key: FOYDMXZTPAEILY-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results