accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 737-31-5

NaCH3OHNINHCH3OIOOI

CAS RN 737-31-5

IUPAC Name: sodium 3,5-diacetamido-2,4,6-triiodobenzoate
Synonyms: sodium amidotrizoate 泛影酸钠 amidotrizoate de sodium natrii amidotrizoas amidotrizoato sodico sodium diacetyldiaminetriiodobenzoate
Molecular Weight (g/mol): 635.9
Molecular Formula: C11H8I3N2NaO4
InChi Key: ZEYOIOAKZLALAP-UHFFFAOYSA-M
SMILES: [Na+].CC(=O)NC1=C(I)C(C([O-])=O)=C(I)C(NC(C)=O)=C1I
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)