accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 73713-79-8

IUPAC Name: 2,1,3-benzothiadiazole-4-sulfonyl chloride
Synonyms: 2,1,3-benzothiadiazole-4-sulfonyl chloride
Molecular Weight (g/mol): 234.67
Molecular Formula: C6H3ClN2O2S2
InChi Key: CXAICGCTHOWKPP-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=CC2=NSN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results