accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHBHOFFFFFF

CAS RN 73852-19-4

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl]boronic acid
Synonyms: 3,5-bis(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 257.93
Molecular Formula: C8H5BF6O2
InChi Key: BPTABBGLHGBJQR-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results