accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HBrBrN

CAS RN 73870-24-3

IUPAC Name: hydrogen 4-(bromomethyl)pyridine bromide
Synonyms: hydrogen 4-(bromomethyl)pyridine bromide
Molecular Weight (g/mol): 252.94
Molecular Formula: C6H7Br2N
InChi Key: VAJUUDUWDNCECT-UHFFFAOYSA-N
SMILES: [H+].[Br-].BrCC1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results