CAS RN 7392-19-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Limetol, TRC
CAS: 7392-19-0 Molecular Formula: C10 H18 O Molecular Weight (g/mol): 154.25 Synonym: 2H-Pyran, 2-ethenyltetrahydro-2,6,6-trimethyl-,2H-Pyran, tetrahydro-2,2,6-trimethyl-6-vinyl- (8CI),Pyran, tetrahydro-2,2,6-trimethyl-6-vinyl- (7CI),2-Ethenyltetrahydro-2,6,6-trimethyl-2H-pyran,2,2,6-Trimethyl-6-ethenyltetrahydrofuran,2,2,6-Trimethyl-6-vinyltetrahydropyran,2,2,6-Trimethyl-6-vinyltetrahydropyrane,2,6,6-Trimethyl-2-vinyltetrahydro-2H-pyran,2,6,6-Trimethyl-2-vinyltetrahydropyran,Bois de Rose oxide,Limetol IUPAC Name: 2-ethenyl-2,6,6-trimethyloxane SMILES: CC1(C)CCCC(C)(O1)C=C