accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HNHN

CAS RN 74-31-7

IUPAC Name: N1,N4-diphenylbenzene-1,4-diamine
Synonyms: DFFD agerite dppd N,N'-diphenyl-P-benzenediamine DPPD permanax 18 antigene P stabilizer dppd antioxidant H permanax dppd usaf GY-2
Molecular Weight (g/mol): 260.34
Molecular Formula: C18H16N2
InChi Key: UTGQNNCQYDRXCH-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC=C(NC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)