CAS RN 74-87-3
IUPAC Name:
chloromethane
Synonyms:
methyl chloride caswell no. 557 artic clorometano MeCl methylchlorid chloor-methaan C-H3-cl cloruro di metile rcra waste number U045
Molecular Weight (g/mol):
50.49
Molecular Formula:
CH3Cl
InChi Key:
NEHMKBQYUWJMIP-UHFFFAOYSA-N
SMILES:
CCl
1
–
4
of
4
results
2
Chloromethane 100 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality