CAS RN 74-99-7
IUPAC Name:
prop-1-yne
Synonyms:
propyne allylene methyl acetylene (substance) propine
Molecular Weight (g/mol):
40.07
Molecular Formula:
C3H4
InChi Key:
MWWATHDPGQKSAR-UHFFFAOYSA-N
SMILES:
CC#C
1
–
2
of
2
results
1
Propyne, ca. 4% in N,N-Dimethylformamide, ca. 1.0mol/L, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Propyne, ca. 4% in N,N-Dimethylformamide, ca. 1.0mol/L, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.