accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CHNOOCH3

CAS RN 7403-41-0

IUPAC Name: N-ethyl-4-methoxybenzamide
Synonyms: benzamide, N-ethyl-4-methoxy-
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: KIOWCVBPJXEJLN-UHFFFAOYSA-N
SMILES: CCNC(=O)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results