accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 74036-95-6

H3CCH3Br

CAS RN 74036-95-6

IUPAC Name: 2-bromotetradecane
Synonyms: 2-bromotetradecane
Molecular Weight (g/mol): 277.29
Molecular Formula: C14H29Br
InChi Key: WZGQIDWFBFDMLE-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCC(C)Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)