accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FONNOHNO

CAS RN 74050-98-9

IUPAC Name: 3-{2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl}-1,2,3,4-tetrahydroquinazoline-2,4-dione
Synonyms: ketanserin ketanserine ketanserina ketaserin ketanserinum
Molecular Weight (g/mol): 395.43
Molecular Formula: C22H22FN3O3
InChi Key: FPCCSQOGAWCVBH-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C1CCN(CCN2C(=O)NC3=CC=CC=C3C2=O)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)