accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 74069-74-2

CAS RN 74069-74-2

IUPAC Name: nonan-2-amine
Synonyms: nonan-2-amine
Molecular Weight (g/mol): 143.27
Molecular Formula: C9H21N
InChi Key: ALXIFCUEJWCQQL-UHFFFAOYNA-N
SMILES: CCCCCCCC(C)N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results