accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7409-09-8

CH3NHOHO

CAS RN 7409-09-8

IUPAC Name: 4-(ethylamino)benzoic acid
Synonyms: 4-(ethylamino)benzoic acid
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: SWXFMMWYVSYQGF-UHFFFAOYSA-N
SMILES: CCNC1=CC=C(C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results