accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrNHNBrN

CAS RN 7411-23-6

IUPAC Name: 3,5-dibromo-1H-1,2,4-triazole
Synonyms: 3,5-dibromo-1H-1,2,4-triazole
Molecular Weight (g/mol): 226.86
Molecular Formula: C2HBr2N3
InChi Key: FRAKFBWDPXYIQO-UHFFFAOYSA-N
SMILES: BrC1=NC(Br)=NN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results