accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3CCH3H3CHNOOOOOOH3CH3CHNH3CCH3

CAS RN 74113-77-2

IUPAC Name: 3,3-bis(2,2,6,6-tetramethylpiperidin-4-yl) 1,5-dioxospiro[5.5]undeca-7,9-diene-3,3-dicarboxylate
Synonyms: 3,3-bis(2,2,6,6-tetramethylpiperidin-4-yl) 1,5-dioxospiro[5.5]undeca-7,9-diene-3,3-dicarboxylate
Molecular Weight (g/mol): 542.72
Molecular Formula: C31H46N2O6
InChi Key: IIZYNJMWYJKDJN-UHFFFAOYSA-N
SMILES: CC1(C)CC(CC(C)(C)N1)OC(=O)C1(CC(=O)C2(CC=CC=C2)C(=O)C1)C(=O)OC1CC(C)(C)NC(C)(C)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)