accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7415-69-2

IUPAC Name: disodium hydrogen ({[(3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms: disodium hydrogen {[(3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxyphosphonate
Molecular Weight (g/mol): 487.17
Molecular Formula: C10H13N5Na2O11P2
InChi Key: LTZCGDIGAHOTKN-JTPGBVSTSA-L
SMILES: [Na+].[Na+].NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1OC(COP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]1O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results