accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7417-21-2

IUPAC Name: 2-(3,4-dimethoxyphenyl)ethan-1-ol
Synonyms: 2-(3,4-dimethoxyphenyl)ethanol
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: SRQAJMUHZROVHW-UHFFFAOYSA-N
SMILES: COC1=CC=C(CCO)C=C1OC
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)