accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 74181-34-3

IUPAC Name: 2,2-dimethyl-1,3-dioxan-5-one
Synonyms: 2,2-dimethyl-1,3-dioxan-5-one
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: ASFQDNDZFGFMMP-UHFFFAOYSA-N
SMILES: CC1(C)OCC(=O)CO1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)