accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClNH2OOH

CAS RN 7424-00-2

IUPAC Name: 1-carboxy-2-(4-chlorophenyl)ethan-1-aminium chloride
Synonyms: fenclonine
Molecular Weight (g/mol): 236.09
Molecular Formula: C9H11Cl2NO2
InChi Key: PFOCEDBJFKVRHU-UHFFFAOYNA-N
SMILES: [Cl-].[NH3+]C(CC1=CC=C(Cl)C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)