accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7424-91-1

CH3OOOH3COH3C

CAS RN 7424-91-1

IUPAC Name: methyl 3,3-dimethoxypropanoate
Synonyms: methyl 3,3-dimethoxypropanoate
Molecular Weight (g/mol): 148.16
Molecular Formula: C6H12O4
InChi Key: SMCVPMKCDDNUCQ-UHFFFAOYSA-N
SMILES: COC(CC(=O)OC)OC
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)