CAS RN 74341-63-2
CAS RN 74341-63-2
IUPAC Name:
2-amino-3-(5-methyl-3-oxo-2,3-dihydro-1,2-oxazol-4-yl)propanoic acid
Synonyms:
AMPA
Molecular Weight (g/mol):
186.17
Molecular Formula:
C7H10N2O4
InChi Key:
UUDAMDVQRQNNHZ-UHFFFAOYNA-N
SMILES:
CC1=C(CC(N)C(O)=O)C(=O)NO1