CAS RN 7439-91-0
Lanthanum pieces, 99.9% (REO)
CAS: 7439-91-0 Molecular Formula: La Molecular Weight (g/mol): 138.91 MDL Number: MFCD00011066 InChI Key: FZLIPJUXYLNCLC-UHFFFAOYSA-N Synonym: in chloride,ion 2,lanthanum, ion la2,atom,ingot,pieces,lanthanum, pieces,2+ ion,foil, 3n,ii cation PubChem CID: 23926 ChEBI: CHEBI:33336 IUPAC Name: lanthanum SMILES: [La]
Lanthanum powder, -40 mesh, 99.7% (REO)
CAS: 7439-91-0 Molecular Formula: La Molecular Weight (g/mol): 138.91 MDL Number: MFCD00011066 InChI Key: FZLIPJUXYLNCLC-UHFFFAOYSA-N Synonym: in chloride,ion 2,lanthanum, ion la2,atom,ingot,pieces,lanthanum, pieces,2+ ion,foil, 3n,ii cation PubChem CID: 23926 ChEBI: CHEBI:33336 IUPAC Name: lanthanum SMILES: [La]
Lanthanum foil, 1.0mm (0.04in) thick, 99.9% (REO)
CAS: 7439-91-0 Molecular Formula: La Molecular Weight (g/mol): 138.91 MDL Number: MFCD00011066 InChI Key: FZLIPJUXYLNCLC-UHFFFAOYSA-N Synonym: in chloride,ion 2,lanthanum, ion la2,atom,ingot,pieces,lanthanum, pieces,2+ ion,foil, 3n,ii cation PubChem CID: 23926 ChEBI: CHEBI:33336 IUPAC Name: lanthanum SMILES: [La]
Lanthanum powder, -200 mesh, 99.9% (REO)
CAS: 7439-91-0 Molecular Formula: La Molecular Weight (g/mol): 138.91 MDL Number: MFCD00011066 InChI Key: FZLIPJUXYLNCLC-UHFFFAOYSA-N Synonym: in chloride,ion 2,lanthanum, ion la2,atom,ingot,pieces,lanthanum, pieces,2+ ion,foil, 3n,ii cation PubChem CID: 23926 ChEBI: CHEBI:33336 IUPAC Name: lanthanum SMILES: [La]
Lanthanum foil, 0.1mm (0.004 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7439-91-0 Molecular Formula: La Molecular Weight (g/mol): 138.91 MDL Number: MFCD00011066 InChI Key: FZLIPJUXYLNCLC-UHFFFAOYSA-N Synonym: in chloride,ion 2,lanthanum, ion la2,atom,ingot,pieces,lanthanum, pieces,2+ ion,foil, 3n,ii cation PubChem CID: 23926 ChEBI: CHEBI:33336 IUPAC Name: lanthanum SMILES: [La]
Lanthanum rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)
CAS: 7439-91-0 Molecular Formula: La Molecular Weight (g/mol): 138.91 MDL Number: MFCD00011066 InChI Key: FZLIPJUXYLNCLC-UHFFFAOYSA-N Synonym: in chloride,ion 2,lanthanum, ion la2,atom,ingot,pieces,lanthanum, pieces,2+ ion,foil, 3n,ii cation PubChem CID: 23926 ChEBI: CHEBI:33336 IUPAC Name: lanthanum SMILES: [La]
Lanthanum rod, 6.35mm (0.25in) dia, 99.9% (REO)
CAS: 7439-91-0 Molecular Formula: La Molecular Weight (g/mol): 138.91 MDL Number: MFCD00011066 InChI Key: FZLIPJUXYLNCLC-UHFFFAOYSA-N Synonym: in chloride,ion 2,lanthanum, ion la2,atom,ingot,pieces,lanthanum, pieces,2+ ion,foil, 3n,ii cation PubChem CID: 23926 ChEBI: CHEBI:33336 IUPAC Name: lanthanum SMILES: [La]
Lanthanum Standard: La @ 10000 μg/mL in 5% HNO3
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lanthanum Standard: La @ 1000 μg/g in Hydrocarbon Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lanthanum Standard, 10,000μg/mL in 2-5% HNO3 (Plasma Grade), SPEX CertiPrep™
CAS: 7439-91-0 Molecular Formula: La Molecular Weight (g/mol): 138.91 MDL Number: MFCD00011066 InChI Key: FZLIPJUXYLNCLC-UHFFFAOYSA-N PubChem CID: 23926 ChEBI: CHEBI:33336 IUPAC Name: lanthanum SMILES: [La]
Lanthanum Standard, 1000μg/mL in 2-5% HNO3 (Assurance Grade), SPEX CertiPrep™
CAS: 7439-91-0 Molecular Formula: La Molecular Weight (g/mol): 138.91 MDL Number: MFCD00011066 InChI Key: FZLIPJUXYLNCLC-UHFFFAOYSA-N PubChem CID: 23926 ChEBI: CHEBI:33336 IUPAC Name: lanthanum SMILES: [La]
Lanthanum Standard: La @ 5000 μg/g in Hydrocarbon Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lanthanum Chloride Releasing Agent: 1% La (from oxide) in 2% HCl, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lanthanum Standard: La @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Lanthanum Nitrate Releasing Agent: 1% La (from oxide) in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
- 1
- 2