CAS RN 7439-92-1
Lead rod, 6.35mm (0.25in) dia, 99.999% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead shot, 3mm (0.1in), 99.999% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead, AAS standard solution, Specpure™ Pb 1000μg/mL
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead powder, -200 mesh, 99% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead foil, 0.1mm (0.004in) thick, Puratronic™, 99.998% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead, 99+%, granular
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead foil, 1.0mm (0.04in) thick, Puratronic™, 99.998% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead powder, -200 mesh, 99.9% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead shot, 4mm (0.2in), 97% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead wire, 2.0mm (0.08in) dia, 99.9% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead rod, 12.7mm (0.5in) dia, Puratronic™, 99.9998% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead rod, 5mm (0.2in) dia, Puratronic™, 99.9999% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead foil, 1.19mm (0.0469in) thick, 99.8% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead rod, 10mm (0.39in) dia, 99.999% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]
Lead foil, 1.6mm (0.063in) thick, 30x30cm, 99+% (metals basis)
CAS: 7439-92-1 Molecular Formula: Pb Molecular Weight (g/mol): 207.20 MDL Number: MFCD00134050 InChI Key: WABPQHHGFIMREM-UHFFFAOYSA-N Synonym: plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant PubChem CID: 5352425 ChEBI: CHEBI:27889 IUPAC Name: lead SMILES: [Pb]