CAS RN 7440-00-8
Neodymium foil, 0.1mm (0.004in) thick, 99.9% (REO)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium rod, 6.35mm (0.25in) dia, 99.5% (metals basis excluding Ta)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium ingot, 99.1% (metals basis)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium powder, -200 mesh, 99.9% (REO)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium powder, -40 mesh, 99.8% (REO)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium foil, 0.3mm (0.012in) thick, 99.9% (REO)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium foil, 1.0mm (0.04in) thick, 99.9% (REO)
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N Synonym: neodimio,neodyme,neodym,trihydride,ingot reo,neodymium, ion nd5+,atom,ingot,iii,foil, 3n PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium Standard: Nd @ 10000 μg/mL in 5% HNO3
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Neodymium @ 5000 μg/g in Hydrocarbon Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Neodymium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]
Neodymium @ 1000 μg/g in Hydrocarbon Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Neodymium Standard: Nd @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Neodymium standard solution, for AAS, 1mg/mL Nd in 2% HNO3, Thermo Scientific™
CAS: 7440-00-8 Molecular Formula: Nd Molecular Weight (g/mol): 144.24 MDL Number: MFCD00011130 InChI Key: QEFYFXOXNSNQGX-UHFFFAOYSA-N PubChem CID: 23934 ChEBI: CHEBI:33372 IUPAC Name: neodymium SMILES: [Nd]