accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7440-09-7

IUPAC Name: potassium(1+) ion
Synonyms: potassium(1+) ion potassium ion potassium(1+)
Molecular Weight (g/mol): 39.1
Molecular Formula: K
InChi Key: NPYPAHLBTDXSSS-UHFFFAOYSA-N
SMILES: [K+]
Grade 
  • (1)
  • (2)
  • (1)
  • (2)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
  • (2)
Industry Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results