CAS RN 7440-10-0
Praseodymium rod, 6.35mm (0.25in) dia, 98.5% (metals basis excluding Ta)
CAS: 7440-10-0 Molecular Formula: Pr Molecular Weight (g/mol): 140.91 MDL Number: MFCD00011174 InChI Key: PUDIUYLPXJFUGB-UHFFFAOYSA-N Synonym: unii-nkn7eza750,nkn7eza750,ingot,praseodimio,praseodyme,praseodym,atom,chips,powder,acmc-1bktg PubChem CID: 23942 ChEBI: CHEBI:49828 IUPAC Name: praseodymium SMILES: [Pr]
Praseodymium foil, 0.1mm (0.004in) thick, 99% (REO)
CAS: 7440-10-0 Molecular Formula: Pr Molecular Weight (g/mol): 140.91 MDL Number: MFCD00011174 InChI Key: PUDIUYLPXJFUGB-UHFFFAOYSA-N Synonym: unii-nkn7eza750,nkn7eza750,ingot,praseodimio,praseodyme,praseodym,atom,chips,powder,acmc-1bktg PubChem CID: 23942 ChEBI: CHEBI:49828 IUPAC Name: praseodymium SMILES: [Pr]
Praseodymium powder, -200 mesh, 99.9% (REO)
CAS: 7440-10-0 Molecular Formula: Pr Molecular Weight (g/mol): 140.91 MDL Number: MFCD00011174 InChI Key: PUDIUYLPXJFUGB-UHFFFAOYSA-N Synonym: unii-nkn7eza750,nkn7eza750,ingot,praseodimio,praseodyme,praseodym,atom,chips,powder,acmc-1bktg PubChem CID: 23942 ChEBI: CHEBI:49828 IUPAC Name: praseodymium SMILES: [Pr]
Praseodymium rod, 12.7mm (0.5in) dia, 99.5% (metals basis excluding Ta)
CAS: 7440-10-0 Molecular Formula: Pr Molecular Weight (g/mol): 140.91 MDL Number: MFCD00011174 InChI Key: PUDIUYLPXJFUGB-UHFFFAOYSA-N Synonym: unii-nkn7eza750,nkn7eza750,ingot,praseodimio,praseodyme,praseodym,atom,chips,powder,acmc-1bktg PubChem CID: 23942 ChEBI: CHEBI:49828 IUPAC Name: praseodymium SMILES: [Pr]
Praseodymium foil, 0.62mm (0.024 in.) thick, 99.5% (REO), Thermo Scientific Chemicals
CAS: 7440-10-0 Molecular Formula: Pr Molecular Weight (g/mol): 140.91 MDL Number: MFCD00011174 InChI Key: PUDIUYLPXJFUGB-UHFFFAOYSA-N Synonym: unii-nkn7eza750,nkn7eza750,ingot,praseodimio,praseodyme,praseodym,atom,chips,powder,acmc-1bktg PubChem CID: 23942 ChEBI: CHEBI:49828 IUPAC Name: praseodymium SMILES: [Pr]
Praseodymium ingot, 99% (REO)
CAS: 7440-10-0 Molecular Formula: Pr Molecular Weight (g/mol): 140.91 MDL Number: MFCD00011174 InChI Key: PUDIUYLPXJFUGB-UHFFFAOYSA-N Synonym: unii-nkn7eza750,nkn7eza750,ingot,praseodimio,praseodyme,praseodym,atom,chips,powder,acmc-1bktg PubChem CID: 23942 ChEBI: CHEBI:49828 IUPAC Name: praseodymium SMILES: [Pr]
Praseodymium foil, 0.25mm (0.01 in.) thick, 99.5% (REO), Thermo Scientific Chemicals
CAS: 7440-10-0 Molecular Formula: Pr Molecular Weight (g/mol): 140.91 MDL Number: MFCD00011174 InChI Key: PUDIUYLPXJFUGB-UHFFFAOYSA-N Synonym: unii-nkn7eza750,nkn7eza750,ingot,praseodimio,praseodyme,praseodym,atom,chips,powder,acmc-1bktg PubChem CID: 23942 ChEBI: CHEBI:49828 IUPAC Name: praseodymium SMILES: [Pr]
Praseodymium foil, 1.0mm (0.04in) thick, 99.5% (REO)
CAS: 7440-10-0 Molecular Formula: Pr Molecular Weight (g/mol): 140.91 MDL Number: MFCD00011174 InChI Key: PUDIUYLPXJFUGB-UHFFFAOYSA-N Synonym: unii-nkn7eza750,nkn7eza750,ingot,praseodimio,praseodyme,praseodym,atom,chips,powder,acmc-1bktg PubChem CID: 23942 ChEBI: CHEBI:49828 IUPAC Name: praseodymium SMILES: [Pr]
Praseodymium Standard: Pr @ 10000 μg/mL in 5% HNO3
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Praseodymium Standard: Pr @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Praseodymium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-10-0 Molecular Formula: Pr Molecular Weight (g/mol): 140.91 MDL Number: MFCD00011174 InChI Key: PUDIUYLPXJFUGB-UHFFFAOYSA-N PubChem CID: 23942 ChEBI: CHEBI:49828 IUPAC Name: praseodymium SMILES: [Pr]