CAS RN 7440-16-6
Rhodium, plasma standard solution, Specpure™ Rh 1000μg/mL
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium wire, 1.0mm (0.04in) dia, 99.8% (metals basis)
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium, 5% on carbon, dry
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium for AAS 1g/L HCl matrix, Fisher chemical™
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium wire, 0.25mm (0.01in) dia, 99.8% (metals basis)
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium black, 99.9% (metals basis)
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium, 1% on alumina powder, reduced
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium 10%, Palladium 1% on activated carbon powder, Thermo Scientific Chemicals
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium wire, 0.127mm (0.005in) dia, 99.8% (metals basis)
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium wire, 0.5mm (0.02in) dia, 99.8% (metals basis)
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium on activated carbon, 5% Rh, unreduced, dry
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium foil, 0.025mm (0.001in) thick, 99.8% (metals basis)
CAS: 7440-16-6
Rhodium, 5% on activated carbon paste, C101023-5
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium powder, -60 mesh, 99.95% (metals basis)
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]