CAS RN 7440-21-3
Silicon lump, 0.1-2.5cm (0.04-1.0in), 99.9999% (metals basis)
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]
Silicon powder, crystalline, APS ∣<50nm, 98%, Laser synthesized from vapor phase
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]
Silicon powder, crystalline, -100+200 mesh, 99.99% (metals basis)
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]
Silicon powder, crystalline, -100 mesh, 99.9% (metals basis)
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]
Silicon powder, APS 1-5 micron, 99.9% (metals basis)
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]
Silicon, 99+%, powder, -325 mesh
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]
Silicon slugs, 9.5mm (0.375 in.) dia. x 9.5mm (0.375 in.) length, 99.9999% (metals basis), Thermo Scientific Chemicals
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]
Silicon granular, virgin poly fines, 99.999% (metals basis)
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]
Silicon powder, crystalline, -325 mesh, 99.5% (metals basis)
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]
Silicon plate, 6.35mm (0.25in) thick, 99.999% (metals basis)
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]
Silicon powder, crystalline, -325 mesh, 99.999% (metals basis)
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]
Silicon lump, 10cm (3.9in) & down, 98.4% (metals basis)
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]
Silicon plate, 9.5mm (0.37in) thick, 99.999% (metals basis)
CAS: 7440-21-3 Molecular Formula: Si Molecular Weight (g/mol): 28.09 MDL Number: MFCD00085311 InChI Key: XUIMIQQOPSSXEZ-UHFFFAOYSA-N Synonym: metal,silicone,defoamer s-10,flots 100sco,powder,dust,unii-z4152n8iui,polyeristalline powder,ccris 6599,ccris 6831 PubChem CID: 5461123 ChEBI: CHEBI:27573 IUPAC Name: silicon SMILES: [Si]