CAS RN 7440-24-6
Strontium for AAS 1g/L HNO3 matrix, Fisher Chemical™
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Strontium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Strontium granules, 19mm (0.76in) & down, 99% (metals basis)
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 MDL Number: MFCD00134060 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N Synonym: strontium, elemental,unii-yzs2rpe8le,yzs2rpe8le,estroncio,granules, 19mm 0.76in & down metals basis,strontium, distilled dendritic pieces,atom,pieces,strontium, chunks,pieces, dendritic PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Strontium granules, 20mm max, 99.0% (metals basis)
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 MDL Number: MFCD00134060 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N Synonym: strontium, elemental,unii-yzs2rpe8le,yzs2rpe8le,estroncio,granules, 19mm 0.76in & down metals basis,strontium, distilled dendritic pieces,atom,pieces,strontium, chunks,pieces, dendritic PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Strontium solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Strontium for AAS 1g/L HCl matrix, Fisher chemical™
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Strontium AA Standard: Sr @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Strontium Standard: Sr @ 10 μg/mL in 2% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Strontium Standard, 1000mg/L in 2% HNO3, (Assurance Grade), SPEX CertiPrep
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Ion Chromatography Standard: Strontium @ 100 μg/mL in H2O, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Strontium Isotopic Standard: Sr @ 100 μg/mL in 2% HNO3 (Natural abundance), VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Strontium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
- 1
- 2