CAS RN 7440-27-9
Terbium foil, 0.1mm (0.004in) thick, 99.9% (REO)
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium pieces, 99.9% (REO)
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium rod, 12.7mm (0.50in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium foil, 0.62mm (0.024in) thick, 99.9% (REO)
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium pieces, distilled dendritic, 99.9% (REO)
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium ingot, 99.9% (REO), Ta <0.4%
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium Standard: Tb @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Terbium Standard: Tb @ 10 μg/mL in 2% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Terbium Standard: Tb @ 10000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Terbium Internal Standard: Tb @ 10 μg/mL in 2% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Terbium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Detector Calibration Multi-Element Mix: Tb, U @ 5; Y, In, Ce, Tl @ 10; Co @ 20; Mg, Sc @ 25; 7Li @ 50; Be @ 100 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
ICP-MS Internal Standard: Tb @ 100 ug/mL in 2% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
- 1
- 2