CAS RN 7440-28-0
Thallium rod, 12.7mm (0.5in) dia, 99.99% (metals basis)
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N Synonym: ramor,thallium, metallic,thallium, elemental,unii-ad84r52xlf,thallium, soluble compounds,ad84r52xlf,thallium, elemental and soluble compounds,rod, 12.7mm 0.5in dia,granules,thallium PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
Thallium rod, 12.7mm (0.5in) dia, 99.999% (metals basis)
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N Synonym: ramor,thallium, metallic,thallium, elemental,unii-ad84r52xlf,thallium, soluble compounds,ad84r52xlf,thallium, elemental and soluble compounds,rod, 12.7mm 0.5in dia,granules,thallium PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
Titanium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
Thallium for AAS 1g/L HNO3 matrix, Fisher chemical™
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N Synonym: ramor,thallium, metallic,thallium, elemental,unii-ad84r52xlf,thallium, soluble compounds,ad84r52xlf,thallium, elemental and soluble compounds,rod, 12.7mm 0.5in dia,granules,thallium PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
ICH/USP Oral Target Elements Standard B: Ni @ 200; Ag, Se @ 150; V @ 100; Co @ 50; Tl @ 8 μg/mL in 2% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
ICH/USP Parenteral Combined-1 Target Elements: Mo @ 1500, Cr @ 1100, Ba @ 700, Sn @ 600, Cu @ 300, Li @ 250, Sb @ 90, Se @ 80, Ag @ 10, Tl @ 8 μg/mL in 5% HNO3, tr. HF, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Thallium Standard: Tl @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Thallium Standard: Tl @ 10000 μg/mL in 5% HNO3
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Thallium AA Standard: Tl @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Sulfur-Free Copper Concentrate: Cu @ approx. 3 wt% in Hydrocarbons, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Sulfur-Free Thallium Concentrate: Tl @ approx. 3 wt% in Hydrocarbons, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Thallium Standard: Tl @ 1000 μg/g in Hydrocarbon Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
- 1
- 2