accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Th

CAS RN 7440-29-1

IUPAC Name: thorium
Synonyms: thorium torio
Molecular Weight (g/mol): 232.04
Molecular Formula: Th
InChi Key: ZSLUVFAKFWKJRC-UHFFFAOYSA-N
SMILES: [Th]
Solvent or Matrix 
  • (7)
Concentration 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results