CAS RN 7440-36-0
Antimony shot, 1-3mm (0.04-0.1in), Puratronic™, 99.9999% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 IUPAC Name: antimony SMILES: [Sb]
Antimony shot, 6mm (0.2in) & down, 99.999% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 IUPAC Name: antimony SMILES: [Sb]
Antimony pieces, 5mm (0.2in) & down, 99.9999% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 IUPAC Name: antimony SMILES: [Sb]
Antimony powder, -200 mesh, 99.999% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 IUPAC Name: antimony SMILES: [Sb]
Antimony powder, -200 mesh, 99.5% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 SMILES: [Sb]
Antimony lump, polycrystalline, Puratronic™, 99.9999% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 SMILES: [Sb]
Antimony broken rod, 12.7mm (0.50in) dia x various lengths, 99.8% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 SMILES: [Sb]
Antimony shot, 1.5-5mm (0.06-0.2in), Puratronic™, 99.9999% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 SMILES: [Sb]
Antimony shot, spherical, 1-5mm (0.04-0.20in), 99.999% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 IUPAC Name: antimony SMILES: [Sb]
Antimony powder, -100 mesh, 99.5% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 IUPAC Name: antimony SMILES: [Sb]
Antimony lump, 1.25cm (0.49in) & down, 99.5% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 SMILES: [Sb]
Antimony rod, 12.7mm (0.50in) dia x 10cm (3.9in) long, 99.8% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 IUPAC Name: antimony SMILES: [Sb]
Antimony shot, 6.35mm (0.25in) & down, 99% (metals basis)
CAS: 7440-36-0 Molecular Formula: Sb Molecular Weight (g/mol): 121.76 MDL Number: MFCD00011211 MFCD00134030 InChI Key: WATWJIUSRGPENY-UHFFFAOYSA-N Synonym: powder,stibium metallicum,antimony, metallic,antimony, elemental,stibanylidyne,unii-9it35j3uv3,antymonowodor polish,unii-0vkz97k3ub,0vkz97k3ub,antimonwasserstoffes german PubChem CID: 5354495 ChEBI: CHEBI:30304 SMILES: [Sb]