CAS RN 7440-38-2
Arsenic powder, -20 mesh, 99% (metals basis)
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N Synonym: arsenicals,arsenic, inorganic,arsenic-75,arsen,gray,grey,black,metallic,compounds,colloidal PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic polycrystalline lump, 2-8mm (0.08-0.3in), Puratronic™, 99.99999+% (metals basis)
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N Synonym: arsenicals,arsenic, inorganic,arsenic-75,arsen,gray,grey,black,metallic,compounds,colloidal PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic lump, 99.999% (metals basis)
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N Synonym: arsenicals,arsenic, inorganic,arsenic-75,arsen,gray,grey,black,metallic,compounds,colloidal PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic Polycrystalline lump, 2-8mm (0.08-0.3 in.), Puratronic™, 99.9999% (Metals basis), Thermo Scientific™
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N Synonym: arsenicals,arsenic, inorganic,arsenic-75,arsen,gray,grey,black,metallic,compounds,colloidal PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic standard solution, for AAS, 1 mg/ml As in 2% KOH
CAS: 7440-38-2 | As | 74.92 g/mol
Arsenic, plasma standard solution, Specpure™ As 1000μg/mL
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.92 MDL Number: MFCD00085309 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Arsenic Standard: As @ 10 μg/mL in 2% HNO3
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
MISA Standard 4: As, Be, Bi, Hg, Re, S, Se @ 100 μg/mL in 10% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
CCT/DRC/ORS Multi-Element Mix 1: Cr, Fe, As, Se @ 100 μg/mL in 2% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
ICP-OES Instrument Check Standard 3: K, P, S @ 100 μg/mL; As, La, Li, Mn, Mo, Na, Ni, Sc @ 20 μg/mL in 5% HCl, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Arsenic AA Standard: As @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
ICP-MS CRQL Solution 2: Ag, As, Be, Cd, Co, Mn, Ni, Pb, Tl, V @ 10; Cr, Cu, Sb, Zn @ 20; Se @ 50; Ba @ 100 μg/mL in 5% HNO3, tr. Tartaric Acid, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Arsenic Standard: As @ 10000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
- 1
- 2