CAS RN 7440-39-3
Barium pieces, 2.5cm (1in) & down, 99.2% (metals basis), Sr ∣<0.8%
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N Synonym: bario,baryum,bario spanish,baryum french,barium, soluble compounds,unii-24gp945v5t,and soluble compounds,barium, elemental,and compounds,atom PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr ∣<0.8%
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N Synonym: bario,baryum,bario spanish,baryum french,barium, soluble compounds,unii-24gp945v5t,and soluble compounds,barium, elemental,and compounds,atom PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
Barium crystalline dendritic solid, 99.9% (metals basis)
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N Synonym: bario,baryum,bario spanish,baryum french,barium, soluble compounds,unii-24gp945v5t,and soluble compounds,barium, elemental,and compounds,atom PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
Barium, 99+%, rods, approx. 2 cm diameter
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N Synonym: bario,baryum,bario spanish,baryum french,barium, soluble compounds,unii-24gp945v5t,and soluble compounds,barium, elemental,and compounds,atom PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
MA6: B, Ba, Ca, Mg, P, Zn @ 750 μg/g in 75 cSt Oil
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
MA6: B, Ba, Ca, Mg, P, Zn @ 1250 μg/g in 75 cSt Oil
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Barium Standard: Ba @ 10000 μg/mL in 5% HNO3
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Tuning Solution 3 For ICP, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Barium AA Standard: Ba @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Barium Standard: Ba @ 5000 μg/g in Hydrocarbon Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Rare Earth and 'Geo' Elements Standard: Ba, Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Rb, Sc, Sm, Sr, Tb, Th, Tm, U, Y, Yb @ 100 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Metal Additives Standard, MA6: B, Ba, Ca, Mg, P, Zn @ 900 μg/g in Hydrocarbon Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Barium Standard: Ba @ 1000 μg/g in Hydrocarbon Oil, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
ICP-MS Tuning Solution: 7Li, Be, Mg, Co, In, Ba, Ce, Pb, Bi, U @10 ug/L in 2% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
ICS Analytes Sub-B2: Ba, Cr, V @ 50 μg/mL in 20% HCl, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications