CAS RN 7440-41-7
Beryllium foil, 0.5mm (0.02in) thick, 99% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium foil, 0.25mm (0.01in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium lump, irregular, 3-38mm (0.1-1.5in), 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium foil, 0.1mm (0.004in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium foil, 0.1mm (0.004in) thick, 99% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium foil, 0.05mm (0.002in) thick, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-41-7 Molecular Formula: Be Molecular Weight (g/mol): 9.01 MDL Number: MFCD00134032 InChI Key: ATBAMAFKBVZNFJ-UHFFFAOYSA-N Synonym: glucinium,dust,metal,powder,metallic,berilio,beryllium, elemental,glucinum,beryllium-9,and compounds PubChem CID: 5460467 ChEBI: CHEBI:33783 IUPAC Name: beryllium SMILES: [Be]
Beryllium Standard: Be @ 10000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Beryllium AA Standard: Be @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
200.7 Rev. 3.3, 6010A - Mixed Calibration Standard 1: Be @ 50; Mn @ 100; Cd, Zn @ 150; Se @ 200; Pb @ 500 μg/mL in 2% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
ICS Analytes Sub-B1: Be, Co, Cu, Mn @ 50; Ag, Cd, Ni, Pb, Zn @ 100 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
ICP-MS Setup Solution: Be, Ce, Fe, In, Li, Mg, Pb, U @ 1 μg/L in 1% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Analytes C Interference Check Standard For ICP/MS, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material