CAS RN 7440-45-1
Cerium foil, 1.0mm (0.04in) thick, 99.9% (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium ingot, 99.8% min (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium ingot, 99% (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium rod, 12.7mm (0.5in) dia, 99.8% (metals basis excluding Ta)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium foil, 0.25mm (0.01in) thick, 99.9% (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium foil, 0.1mm (0.004in) thick, 99.9% (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium powder, -60 mesh, 99.9% (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium foil, 0.62mm (0.024in) thick, 99.9% (REO)
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N Synonym: cerio,ingot,cerium, chips,foil, 3n,zer,ceramics-aeium,acmc-1bl5r,chips, 3n reo,powder,-200 mesh,cerium PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium, plasma standard solution, Specpure™ Ce 1000μg/mL
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-45-1 Molecular Formula: Ce Molecular Weight (g/mol): 140.12 MDL Number: MFCD00010924 InChI Key: GWXLDORMOJMVQZ-UHFFFAOYSA-N PubChem CID: 23974 ChEBI: CHEBI:33369 IUPAC Name: cerium SMILES: [Ce]
Cerium Standard: Ce @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Cerium Standard: Ce @ 10000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
ICP-MS Tuning/Mass Calibration Multi-Element Mix 1 (concentrate): 7Li, Y, Ce, Tl @ 10 ug/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
- 1
- 2