accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Cs

CAS RN 7440-46-2

IUPAC Name: caesium
Synonyms: caesium cesium cesio caesium atom zaesium
Molecular Weight (g/mol): 132.91
Molecular Formula: Cs
InChi Key: TVFDJXOCXUVLDH-UHFFFAOYSA-N
SMILES: [Cs]
Molecular Weight (g/mol) 
  • (7)
  • (1)
  • (10)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
Solvent or Matrix 
  • (1)
  • (8)
Concentration 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (8)
  • (7)
  • (1)
  • (10)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (8)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)