CAS RN 7440-48-4
Cobalt slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.95% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt powder, -22 mesh, Puratronic™, 99.998% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt powder, -400 mesh, 99.5% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt powder, 1.6 micron, 99.8% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.95% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt wire, 1.5mm (0.06in) dia, 99.95% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt wire, 0.1mm (0.004in) dia, Puratronic™, 99.995% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt foil, 1.5mm (0.06in) thick, 99.95% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt pieces, 12mm (0.47in) & down, Vacuum Deposition Grade, 99.5% (metals basis excluding Ni), Ni nominal 1%
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt rod, 5mm (0.2in) dia, 99.95% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt foil, 0.05mm (0.002in) thick, 99.95% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt powder, spherical, -100+325 mesh, 99.8% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.995% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt wire, 0.5mm (0.02in) dia, Puratronic™, 99.995% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt wire, 2.0mm (0.08in) dia, 99.95% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]