CAS RN 7440-50-8
Copper turnings, 99+% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper gauze, 50 mesh woven from 0.23mm (0.009in) dia wire
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper shot, 3-6mm (0.12-0.24in), Puratronic™, 99.999% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper shot, 0.8-2mm (0.03-0.08in), 99.5% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper shot, 1-10mm (0.04-0.4in), 99.9% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper standard solution, for AAS, 1 mg/ml Cu in 2 to 5% HNO3
CAS: 7440-50-8 | Cu | 63.55 g/mol
Copper, AAS standard solution, Specpure™ Cu 1000μg/mL
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper shot, 6mm (0.24in) & down, Puratronic™, 99.9999% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper powder, -40+100 mesh, 99.5% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper powder, spherical, -100+325 mesh, 99.9% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper powder, -325 mesh, 10% max +325 mesh, 99% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.9999% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper foil, 0.05mm (0.002in) thick, Puratronic™, 99.9999% (metals basis)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper, 99%, powder, max. 106 μm (-140 mesh)
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Copper slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.55 MDL Number: MFCD00010965 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N Synonym: cuprum,cobre,cuivre,blister,cathode,bronze,powder,anode,precipitates,kupfer PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]