CAS RN 7440-52-0
Erbium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium foil, 0.25mm (0.01in) thick, 99.9% (REO)
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium foil, 0.62mm (0.024in) thick, 99.9% (REO)
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium pieces, 99.9% (REO)
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium powder, -40 mesh, 99.9% (REO)
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium foil, 0.025mm (0.001 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7440-52-0 Molecular Formula: Er Molecular Weight (g/mol): 167.26 MDL Number: MFCD00010987 InChI Key: UYAHIZSMUZPPFV-UHFFFAOYSA-N Synonym: unii-77b218d3ye,foil,iii ion,erbio,pieces,powder,chips,ingot,er3,pieces, distilled dendritic PubChem CID: 23980 ChEBI: CHEBI:33379 IUPAC Name: erbium SMILES: [Er]
Erbium Standard: Er @ 10000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Erbium Standard: Er @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications