CAS RN 7440-53-1
Europium ingot, 99.9% (REO)
CAS: 7440-53-1 Molecular Formula: Eu Molecular Weight (g/mol): 151.96 MDL Number: MFCD00010992 InChI Key: OGPBJKLSAFTDLK-UHFFFAOYSA-N Synonym: europio,atom,ingot,acmc-1bj8v,europium, chip, in mineral oil,foil, 1.0mm 0.04in thick,foil, 0.1mm 0.004in thick,foil, 0.25mm 0.01in thick,rod, 9.52 mm 0.375in dia,ingot reo , packed in mineral oil PubChem CID: 23981 ChEBI: CHEBI:32999 IUPAC Name: europium SMILES: [Eu]
Europium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
CAS: 7440-53-1 Molecular Formula: Eu Molecular Weight (g/mol): 151.96 MDL Number: MFCD00010992 InChI Key: OGPBJKLSAFTDLK-UHFFFAOYSA-N PubChem CID: 23981 ChEBI: CHEBI:32999 IUPAC Name: europium SMILES: [Eu]
Europium foil, 0.25mm (0.01in) thick, 99.9% (REO)
CAS: 7440-53-1 Molecular Formula: Eu Molecular Weight (g/mol): 151.96 MDL Number: MFCD00010992 InChI Key: OGPBJKLSAFTDLK-UHFFFAOYSA-N Synonym: europio,atom,ingot,acmc-1bj8v,europium, chip, in mineral oil,foil, 1.0mm 0.04in thick,foil, 0.1mm 0.004in thick,foil, 0.25mm 0.01in thick,rod, 9.52 mm 0.375in dia,ingot reo , packed in mineral oil PubChem CID: 23981 ChEBI: CHEBI:32999 IUPAC Name: europium SMILES: [Eu]
Europium rod, 9.52 mm (0.375in) dia, 99.9% (metals basis)
CAS: 7440-53-1 Molecular Formula: Eu Molecular Weight (g/mol): 151.96 MDL Number: MFCD00010992 InChI Key: OGPBJKLSAFTDLK-UHFFFAOYSA-N Synonym: europio,atom,ingot,acmc-1bj8v,europium, chip, in mineral oil,foil, 1.0mm 0.04in thick,foil, 0.1mm 0.004in thick,foil, 0.25mm 0.01in thick,rod, 9.52 mm 0.375in dia,ingot reo , packed in mineral oil PubChem CID: 23981 ChEBI: CHEBI:32999 IUPAC Name: europium SMILES: [Eu]
Europium for AAS 1g/L HNO3 matrix, Fisher chemical™
CAS: 7440-53-1 Molecular Formula: Eu Molecular Weight (g/mol): 151.96 MDL Number: MFCD00010992 InChI Key: OGPBJKLSAFTDLK-UHFFFAOYSA-N PubChem CID: 23981 ChEBI: CHEBI:32999 IUPAC Name: europium SMILES: [Eu]
Europium Standard: Eu @ 10000 μg/mL in 5% HNO3
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Europium Standard: Eu @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Europium pieces, dendritic, 99.9% (REO)
CAS: 7440-53-1 Molecular Formula: Eu Molecular Weight (g/mol): 151.96 MDL Number: MFCD00010992 InChI Key: OGPBJKLSAFTDLK-UHFFFAOYSA-N Synonym: europio,atom,ingot,acmc-1bj8v,europium, chip, in mineral oil,foil, 1.0mm 0.04in thick,foil, 0.1mm 0.004in thick,foil, 0.25mm 0.01in thick,rod, 9.52 mm 0.375in dia,ingot reo , packed in mineral oil PubChem CID: 23981 ChEBI: CHEBI:32999 IUPAC Name: europium SMILES: [Eu]