CAS RN 7440-60-0
Holmium foil, 0.3mm (0.01in) thick, 99.9% (REO)
CAS: 7440-60-0 Molecular Formula: Ho Molecular Weight (g/mol): 164.93 MDL Number: MFCD00011049 InChI Key: KJZYNXUDTRRSPN-UHFFFAOYSA-N Synonym: atom,unii-w1xx32sqn1,w1xx32sqn1,foil,holmio,chips,ingot,pieces,powder,acmc-1c9ym PubChem CID: 23988 ChEBI: CHEBI:49648 IUPAC Name: holmium SMILES: [Ho]
Holmium pieces, 99.9% (metals basis excluding Ta)
CAS: 7440-60-0 Molecular Formula: Ho Molecular Weight (g/mol): 164.93 MDL Number: MFCD00011049 InChI Key: KJZYNXUDTRRSPN-UHFFFAOYSA-N Synonym: atom,unii-w1xx32sqn1,w1xx32sqn1,foil,holmio,chips,ingot,pieces,powder,acmc-1c9ym PubChem CID: 23988 ChEBI: CHEBI:49648 IUPAC Name: holmium SMILES: [Ho]
Holmium foil, 0.127mm (0.005in) thick, 99.9% (REO)
CAS: 7440-60-0 Molecular Formula: Ho Molecular Weight (g/mol): 164.93 MDL Number: MFCD00011049 InChI Key: KJZYNXUDTRRSPN-UHFFFAOYSA-N Synonym: atom,unii-w1xx32sqn1,w1xx32sqn1,foil,holmio,chips,ingot,pieces,powder,acmc-1c9ym PubChem CID: 23988 ChEBI: CHEBI:49648 IUPAC Name: holmium SMILES: [Ho]
Holmium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
CAS: 7440-60-0 Molecular Formula: Ho Molecular Weight (g/mol): 164.93 MDL Number: MFCD00011049 InChI Key: KJZYNXUDTRRSPN-UHFFFAOYSA-N PubChem CID: 23988 ChEBI: CHEBI:49648 IUPAC Name: holmium SMILES: [Ho]
Holmium pieces, distilled dendritic, 99.9% (REO)
CAS: 7440-60-0 Molecular Formula: Ho Molecular Weight (g/mol): 164.93 MDL Number: MFCD00011049 InChI Key: KJZYNXUDTRRSPN-UHFFFAOYSA-N Synonym: atom,unii-w1xx32sqn1,w1xx32sqn1,foil,holmio,chips,ingot,pieces,powder,acmc-1c9ym PubChem CID: 23988 ChEBI: CHEBI:49648 IUPAC Name: holmium SMILES: [Ho]
Holmium rod, 6.35mm (0.25in) dia, 99.8% (metals basis)
CAS: 7440-60-0 Molecular Formula: Ho Molecular Weight (g/mol): 164.93 MDL Number: MFCD00011049 InChI Key: KJZYNXUDTRRSPN-UHFFFAOYSA-N Synonym: atom,unii-w1xx32sqn1,w1xx32sqn1,foil,holmio,chips,ingot,pieces,powder,acmc-1c9ym PubChem CID: 23988 ChEBI: CHEBI:49648 IUPAC Name: holmium SMILES: [Ho]
Holmium foil, 0.62mm (0.024 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7440-60-0 Molecular Formula: Ho Molecular Weight (g/mol): 164.93 MDL Number: MFCD00011049 InChI Key: KJZYNXUDTRRSPN-UHFFFAOYSA-N Synonym: atom,unii-w1xx32sqn1,w1xx32sqn1,foil,holmio,chips,ingot,pieces,powder,acmc-1c9ym PubChem CID: 23988 ChEBI: CHEBI:49648 IUPAC Name: holmium SMILES: [Ho]
Holmium foil, 0.025mm (0.001in) thick, 99.9% (REO)
CAS: 7440-60-0 Molecular Formula: Ho Molecular Weight (g/mol): 164.93 MDL Number: MFCD00011049 InChI Key: KJZYNXUDTRRSPN-UHFFFAOYSA-N Synonym: atom,unii-w1xx32sqn1,w1xx32sqn1,foil,holmio,chips,ingot,pieces,powder,acmc-1c9ym PubChem CID: 23988 ChEBI: CHEBI:49648 IUPAC Name: holmium SMILES: [Ho]
Holmium foil, 1.0mm (0.04 in.) thick, 99.9% (REO), Thermo Scientific Chemicals
CAS: 7440-60-0 Molecular Formula: Ho Molecular Weight (g/mol): 164.93 MDL Number: MFCD00011049 InChI Key: KJZYNXUDTRRSPN-UHFFFAOYSA-N Synonym: atom,unii-w1xx32sqn1,w1xx32sqn1,foil,holmio,chips,ingot,pieces,powder,acmc-1c9ym PubChem CID: 23988 ChEBI: CHEBI:49648 IUPAC Name: holmium SMILES: [Ho]
Holmium for AAS 1g/L HNO3 matrix, Fisher chemical™
CAS: 7440-60-0 Molecular Formula: Ho Molecular Weight (g/mol): 164.93 MDL Number: MFCD00011049 InChI Key: KJZYNXUDTRRSPN-UHFFFAOYSA-N PubChem CID: 23988 ChEBI: CHEBI:49648 IUPAC Name: holmium SMILES: [Ho]
Holmium powder, -200 mesh, 99.9% (REO)
CAS: 7440-60-0 Molecular Formula: Ho Molecular Weight (g/mol): 164.93 MDL Number: MFCD00011049 InChI Key: KJZYNXUDTRRSPN-UHFFFAOYSA-N Synonym: atom,unii-w1xx32sqn1,w1xx32sqn1,foil,holmio,chips,ingot,pieces,powder,acmc-1c9ym PubChem CID: 23988 ChEBI: CHEBI:49648 IUPAC Name: holmium SMILES: [Ho]
Holmium Standard: Ho @ 10000 μg/mL in 5% HNO3
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
MISA Standard 5: Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb @ 100 μg/mL in 2% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Holmium Standard: Ho @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
- 1
- 2