CAS RN 7440-62-2
Vanadium powder, -325 mesh, 99.5% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium pieces, 99.7% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium rod, 6.35mm (0.25in) dia, 99.5% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium wire, 1.5mm (0.06in) dia, 99.8% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium wire, 2.0mm (0.08 in.) dia., 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium rod, 12.7mm (0.5in) dia, 99.5% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.8% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium wire, 0.075mm (0.003in) dia, 99.8% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium wire, 1.0mm (0.04in) dia, 99.5% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium foil, 0.127mm (0.005in) thick, 99.8% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium granules, 1-3mm (0.04-0.12in), Vacuum Deposition Grade, 99.7% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium wire, 0.25mm (0.01in) dia, 99.8% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium standard solution, for AAS, 1 mg/ml V(3+) in 2% HNO3
CAS: 7440-62-2 | V | 50.94 g/mol
Vanadium slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.8% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]
Vanadium slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.8% (metals basis)
CAS: 7440-62-2 Molecular Formula: V Molecular Weight (g/mol): 50.94 MDL Number: MFCD00011453 InChI Key: LEONUFNNVUYDNQ-UHFFFAOYSA-N Synonym: dust,vanadium, elemental,ion,iii,1+ , ion,vanadium, ion 3+,fume or dust,vanadium, ion v5+,unii-00j9j9xkde,00j9j9xkde PubChem CID: 23990 ChEBI: CHEBI:35170 IUPAC Name: vanadium SMILES: [V]