CAS RN 7440-64-4
Ytterbium pieces, sublimed, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium for AAS 1g/L HNO3 matrix, Fisher chemical™
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium foil, 1.0mm (0.04in) thick, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium foil, 0.1mm (0.004in) thick, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium foil, 0.64mm (0.025in) thick, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium powder, -200 mesh, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium powder, -40 mesh, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium foil, 0.3mm (0.01in) thick, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium ingot, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium Standard: Yb @ 10000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Ytterbium Standard: Yb @ 1000 μg/mL in 5% HNO3, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
- 1
- 2